Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyltransferase like 3 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10077311IR
Description
Methyltransferase like 3 Polyclonal antibody specifically detects Methyltransferase like 3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Methyltransferase like 3 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
adoMet-binding subunit of the human mRNA (N6-adenosine)-methyltransferase, EC 2.1.1.62, IME4, M6A, methyltransferase like 3, Methyltransferase-like protein 3, MGC4336, mRNA m(6)A methyltransferase, MTA70, MT-A70, N6-adenosine-methyltransferase 70 kDa subunit, Spo8 | |
The immunogen recognized by this antibody maps to a region between residue 530 and 580 of human methyltransferase like 3 (N6-adenosine-methyltransferase 70 kDa subunit) using the numbering given in entry NP_062826.2 (GeneID 56339). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
56339 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction