Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyltransferase like 3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10077311
Description
Methyltransferase like 3 Polyclonal specifically detects Methyltransferase like 3 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative).Specifications
Methyltransferase like 3 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
adoMet-binding subunit of the human mRNA (N6-adenosine)-methyltransferase, EC 2.1.1.62, IME4, M6A, methyltransferase like 3, Methyltransferase-like protein 3, MGC4336, mRNA m(6)A methyltransferase, MTA70, MT-A70, N6-adenosine-methyltransferase 70 kDa subunit, Spo8 | |
Rabbit | |
Affinity Purified | |
RUO | |
56339 | |
Store at 4C. Do not freeze. |
Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
1.0 mg/mL | |
Immunohistochemistry, Immunoprecipitation 2 - 5 ug/mg lysate, Immunohistochemistry-Paraffin 1:500- 1:2000, Western Blot (Negative) | |
NP_062826.2 | |
METTL3 | |
The immunogen recognized by this antibody maps to a region between residue 530 and 580 of human methyltransferase like 3 (N6-adenosine-methyltransferase 70 kDa subunit) using the numbering given in entry NP_062826.2 (GeneID 56339). | |
100 μL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction