Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyltransferase like 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $487.50

Specifications

Antigen Methyltransferase like 3
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092496
SDP
View Documents
Novus Biologicals
NBP30329020UL
20 μL
Each for $181.90
Only null left
Add to Cart
 
NB092495
SDP
View Documents
Novus Biologicals
NBP303290100UL
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

Methyltransferase like 3 Polyclonal specifically detects Methyltransferase like 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

Methyltransferase like 3
Polyclonal
Purified
RUO
PBS with 50% glycerol, pH7.3.
56339
IgG
Affinity purified
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Rabbit
Human, Mouse, Rat
adoMet-binding subunit of the human mRNA (N6-adenosine)-methyltransferase, EC 2.1.1.62, IME4, M6A, methyltransferase like 3, Methyltransferase-like protein 3, MGC4336, mRNA m(6)A methyltransferase, MTA70, MT-A70, N6-adenosine-methyltransferase 70 kDa subunit, Spo8
Recombinant fusion protein containing a sequence corresponding to amino acids 1-156 of human Methyltransferase like 3 (NP_062826.2).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.