Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
methyltransferase like 9 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298039PE
Description
methyltransferase like 9 Polyclonal antibody specifically detects methyltransferase like 9 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
methyltransferase like 9 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
DORA reverse strand protein, DORA reverse strand protein 1, DREV, DREV1methyltransferase-like protein 9, FLJ21912, methyltransferase like 9, p53 activated protein 1, PAP1 | |
Produced in rabbits immunized with E. coli-derived Human methyltransferase like 9 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
51108 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction