Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyltriethoxysilane, 98+%, ACROS Organics™

Click to view available options
Quantity:
25mL
250mL
1L
Packaging:
Glass bottle
2031-67-6
C7H18O3Si
178.3
MFCD00009066
CPUDPFPXCZDNGI-UHFFFAOYSA-N
methyltriethoxysilane,triethoxy methyl silane,methyl triethoxysilane,silane, triethoxymethyl,triethoxysilylmethane,union carbide a-162,ici-ep 5850,silane, methyltriethoxy,methaneorthosiliconic acid, triethyl ester,mtes
16241
triethoxy(methyl)silane
CCO[Si](C)(OCC)OCC
2031-67-6
C7H18O3Si
178.3
MFCD00009066
CPUDPFPXCZDNGI-UHFFFAOYSA-N
methyltriethoxysilane,triethoxy methyl silane,methyl triethoxysilane,silane, triethoxymethyl,triethoxysilylmethane,union carbide a-162,ici-ep 5850,silane, methyltriethoxy,methaneorthosiliconic acid, triethyl ester,mtes
16241
triethoxy(methyl)silane
CCO[Si](C)(OCC)OCC
Viscosity 0.6 mPa.s (20°C)
Melting Point -40.0°C
Color Colorless
Density 0.8900g/mL
Flash Point 23°C
Specific Gravity 0.89
Infrared Spectrum Authentic
Quantity 250mL
Packaging Glass bottle
Refractive Index 1.3825 to 1.3845
Boiling Point 141.0°C to 143.0°C
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (GC)
Linear Formula CH3Si(OC2H5)3
Beilstein 04,629
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 178.3
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material Methyltriethoxysilane, 98+%
Show More Show Less

GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g

GHS Signal Word: Warning

EINECSNumber : 217-983-9

RTECSNumber : VV4640000

TSCA : TSCA