Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyltriphenoxyphosphonium iodide, 95%

Catalog No. p-3779250
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC415020050 5 g Glass bottle
AC415020500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC415020050 Supplier Thermo Scientific Chemicals Supplier No. 415020050
Only null left
Add to Cart

CAS: 17579-99-6 | C19H18IO3P | 452.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17579-99-6
Molecular Formula C19H18IO3P
Molecular Weight (g/mol) 452.22
MDL Number MFCD00011911
InChI Key VKTOBGBZBCELGC-UHFFFAOYSA-M
Synonym methyltriphenoxyphosphonium iodide, methyltriphenoxyphosphanium iodide, acmc-20ake2, triphenyl phosphite methoiodide, methyl triphenoxyphosphonium iodide, methyl triphenoxy phosphanium iodide, methyltriphenoxyphosphorus 1+ iodide, phosphorus 1+ ,methyltriphenoxy-, iodide 1:1 , t-4
PubChem CID 2735090
IUPAC Name methyl(triphenoxy)phosphanium;iodide
SMILES C[P+](OC1=CC=CC=C1)(OC2=CC=CC=C2)OC3=CC=CC=C3.[I-]

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 141.0°C to 146.0°C
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Beilstein 06, 177
Fieser 01,1249; 02,446; 04,557; 06,649; 08,354
Solubility Information Solubility in water: negligible. Other solubilities: soluble in acetone,diethyl ether
Formula Weight 452.22
Percent Purity 95%
Chemical Name or Material Methyltriphenoxyphosphonium iodide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 241-551-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.