Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Tolunitrile, 98+%

Catalog No. AC158020010
Click to view available options
Quantity:
5 g
100 g
250 g
1 kg
Packaging:
Glass bottle

CAS: 104-85-8 | C8H7N | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-85-8
Molecular Formula C8H7N
Molecular Weight (g/mol) 117.15
MDL Number MFCD00001827
InChI Key VCZNNAKNUVJVGX-UHFFFAOYSA-N
Synonym p-tolunitrile, 4-cyanotoluene, benzonitrile, 4-methyl, 4-tolunitrile, p-methylbenzonitrile, p-toluonitrile, p-cyanotoluene, p-tolylnitrile, p-tolunitril, p-toluenenitrile
PubChem CID 7724
IUPAC Name 4-methylbenzonitrile
SMILES CC1=CC=C(C=C1)C#N

Specifications

Melting Point 26.0°C to 28.0°C
Density 0.9800g/mL
Boiling Point 103.0°C to 106.0°C (20.0 mmHg)
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5285 to 1.5305
Linear Formula CH3C6H4CN
Quantity 1 kg
Beilstein 09, 489
Specific Gravity 0.98
Merck Index 15, 9698
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 117.15
Percent Purity 98+%
Physical Form Viscous Liquid
Chemical Name or Material p-Tolunitrile, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
EINECSNumber 203-244-8
RTECSNumber XV0700000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.