Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylzinc chloride, 2M solution in THF, AcroSeal™

Catalog No. ac436080500
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 5158-46-3 | CH3ClZn | 115.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5158-46-3, 109-99-9
Molecular Formula CH3ClZn
Molecular Weight (g/mol) 115.87
MDL Number MFCD00192334
InChI Key NMLXKNNXODLJIN-UHFFFAOYSA-M
Synonym methylzinc chloride, chloro methyl zinc, methylzinc chloride solution, methylzinc chloride solution, 2.0 m in thf
PubChem CID 4152418
IUPAC Name carbanide;chlorozinc(1+)
SMILES [CH3-].Cl[Zn+]

Specifications

Color Colorless to Yellow
CAS Min % 21.31, 75.32
CAS Max % 24.68, 78.69
Physical Form Turbid Solution
Quantity 50 mL
Flash Point −17°C
Formula Weight 115.87
Concentration or Composition (by Analyte or Components) 1.90 to 2.20M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.033
Density 1.0330g/mL
Chemical Name or Material Methylzinc chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
May cause respiratory irritation.
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.