Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ METL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558301
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558301 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558301 Supplier Invitrogen™ Supplier No. PA558301
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DIDEDALEIF NRNAEEFELT NIDMVQCDVC LLSNRMSKSF DTVIMNPPFG T Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 84%.

Catalytic subunit of a heterodimer with TRMT112, which specifically methylates the 6th position of adenine in position 1832 of 18S rRNA. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen METL5
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene METTL5
Gene Accession No. Q9NRN9
Gene Alias 2810410A08Rik; DC3; HSPC133; im:7137598; methyltransferase like 5; methyltransferase-like protein 5; Mettl5; RGD1566062; rRNA N6-adenosine-methyltransferase METTL5; rRNA N6-adenosine-methyltransferase METTL5; methyltransferase-like protein 5; zgc:103655
Gene Symbols METTL5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human METL5. Recombinant protein control fragment (Product #RP-96101).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29081
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.