Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ METL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115571
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115571 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115571 Supplier Invitrogen™ Supplier No. PA5115571
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Methyltransferase like 5.

Catalytic subunit of a heterodimer with TRMT112, which specifically methylates the 6th position of adenine in position 1832 of 18S rRNA. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen METL5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene METTL5
Gene Accession No. Q8K1A0, Q9NRN9
Gene Alias 2810410A08Rik; DC3; HSPC133; im:7137598; methyltransferase like 5; methyltransferase-like protein 5; Mettl5; RGD1566062; rRNA N6-adenosine-methyltransferase METTL5; rRNA N6-adenosine-methyltransferase METTL5; methyltransferase-like protein 5; zgc:103655
Gene Symbols METTL5
Host Species Rabbit
Immunogen A synthesized peptide derived from human METTL5(Accession Q9NRN9), corresponding to amino acid residues V105-S155.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29081, 502632, 75422
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.