Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metnase Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501565611
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-5611 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-5611 Supplier Bethyl Laboratories, Inc Supplier No. A303615A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

SETMAR (SET domain and mariner transposase fusion gene) is a histone methyltransferase that methylates Lys-4 and Lys-36 of histone H3, creating specific tags for epigenetic transcriptional activation. It specifically mediates dimethylation of H3 Lys-36. SETMAR has sequence-specific DNA-binding activity, and recognizes the 19-mer core of the 5′-terminal inverted repeats (TIRs) of the Hsmar1 transposable element. SETMAT has DNA nicking activity, and in vivo end-joining activity, and may mediate genomic integration of foreign DNA.

Specifications

Antigen Metnase
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene SETMAR
Gene Accession No. Q53H47
Gene Alias SETMAR; SET domain and mariner transposase fusion gene; METNASE; SET domain and mariner transposase fusion
Gene Symbols SETMAR
Host Species Rabbit
Immunogen Between 225 and 275
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6419
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.