Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metoclopramide Hydrochloride, Monohydrate, USP, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18604554 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle

ME176, 54143-57-6, C14H22ClN3O2·HCI·H2O

Spectrum™ Chemical Metoclopramide Hydrochloride, Monohydrate, USP is a medication used mostly for stomach and esophageal problems to treat and prevent nausea and vomiting. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 54143-57-6
Molecular Formula C14H25Cl2N3O3
Molecular Weight (g/mol) 354.27
MDL Number MFCD00150634
InChI Key KJBLQGHJOCAOJP-UHFFFAOYSA-N
IUPAC Name 4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide hydrate hydrochloride
SMILES O.Cl.CCN(CC)CCNC(=O)C1=CC(Cl)=C(N)C=C1OC

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Quantity 5 kg
Percent Purity 98 to 101%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.