Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Metoprolol tartrate
GREENER_CHOICESDP

Catalog No. 325650 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-565-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 32-565-0 Supplier R&D Systems™ Supplier No. 3256/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective β1 antagonist

Selective β1-adrenoceptor antagonist (Ki values are 47, 2960 and 10100 nM for β1, β2 and β3 adrenoceptors respectively). Inhibits spontaneous endothelin-1 production in vitro and displays antihypertensive and antianginal activity in vivo.

Chemical Identifiers

CAS 56392-17-7
Molecular Formula C34H56N2O12

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 342.4
Purity >99%
Molecular Weight (g/mol) 684.824
Chemical Name or Material Metoprolol tartrate
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.