Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEKISUI Diagnostics Metrix™ COVID-19 Test

Catalog No. 22321140 Shop All SEKISUI Diagnostics Products
Change view
Click to view available options
Quantity:
25 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-321-140 25 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-321-140 Supplier SEKISUI Diagnostics Supplier No. MTRXC1925PK

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

The Metrix™ COVID-19 test includes clinical claims for symptomatic and asymptomatic individuals. It allows for an enhanced point-of-care testing experience with dual-sample types for nasal or saliva.

  • Molecular accuracy without the wait - Improve clinical outcomes by administering immediate treatment in just 30 minutes.
  • Accessible molecular testing - Breakthrough platform designed to eliminate cost-barriers.
  • Reduce repeat testing - No need for serial or confirmation testing when nasal specimen is used.
  • Alleviates testing restrictions - Indicated for symptomatic and asymptomatic testing, allowing for screening prior to medical procedures.
  • Simple workflow - Can be performed in any point-of-care setting, with minimal training required.
  • Convenient sampling - The only test with dual-sample types, allowing you to provide the best patient experience possible.
  • Exceptional Support - Experienced support teams and online training modules to help streamline your testing implementation.
TRUSTED_SUSTAINABILITY

Specifications

CE Marker No
Clia Complexity Waived (FDA, EUA)
Detectable Analytes COVID-19
Type COVID-19 Test
Disposable Reusable
For Use With (Application) Point-of-Care Testing
Includes 25 Tests
Quantity 25 Tests/Kit
Sterility Sterile
Test Time 30 min.
Form Liquid Ready-to-Use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.