Learn More
Invitrogen™ METRNL Polyclonal Antibody

Description
METRNL Polyclonal Antibody for ELISA, Western Blot
Specifications
Specifications
| Antigen | METRNL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 |
| Gene | METRNL |
| Gene Accession No. | Q5RJL6, Q641Q3, Q8VE43 |
| Gene Alias | 9430048M07Rik; BC019776; cometin; meteorin like, glial cell differentiation regulator; meteorin, glial cell differentiation regulator-like; meteorin-like protein; METRNL; Subfatin |
| Gene Symbols | METRNL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.