Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metro™ MetroMax i Glassware Cart, Accessories

Catalog No. p-4597697 Shop All Metro Products
Click to view available options
Product Type:
Basket
Glassware Transport Kit

For use with MetroMax i carts

  • 36 in. basket for glassware cart

Specifications

Product Type Glassware Transport Kit
Color Gray
For Use With (Application) For easy and efficient way to transport laboratory Glassware
For Use With (Equipment) For MetroMax Glassware carts
Includes 6 in. Deep Metrotote box, two long divider, two short divider
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.