Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
METT10D Antibody (OTI3E8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272653H
Description
METT10D Monoclonal specifically detects METT10D in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry.Specifications
METT10D | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
METTL16 | |
Full length recombinant protein of human METT10D (NP_076991) produced in HEK293T cells. | |
0.1 mL | |
79066.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3E8 | |
Western Blot, Immunohistochemistry | |
EC 2.1.1.-, methyltransferase 10 domain containing, Methyltransferase 10 domain-containing protein, methyltransferase like 16, METT10D, MGC3329, putative methyltransferase METT10D | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Canine | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction