Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METT10D Monoclonal Antibody (OTI5F10), TrueMAB™, OriGene
SDP

Catalog No. 502540166
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0166 30 μL
50-167-5328 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0166 Supplier OriGene Supplier No. TA504697S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

METT10D Monoclonal antibody specifically detects METT10D in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen METT10D
Applications Western Blot
Classification Monoclonal
Clone OTI5F10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CPA2
Gene Accession No. P19222, P48052, Q504N0
Gene Alias 2610100D03Rik; 2810013M15Rik; A830095F14Rik; AI846133; methyltransferase 10 domain containing; methyltransferase 10 domain-containing protein; methyltransferase like 16; methyltransferase-like protein 16; METT10D; METTL16; N6-adenosine-methyltransferase METTL16; putative methyltransferase METT10D; RNA N6-adenosine-methyltransferase METTL16; U6 small nuclear RNA (adenine-(43)-N(6))-methyltransferase
Host Species Mouse
Immunogen Full length human recombinant protein of human CPA2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1358, 232680, 296959
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.