Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ METT10D Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554185
Description
Immunogen sequence: DERSEEKGGV EVLESCQGSS NGAQDQEASE QFGSPVAERG KRLPGVAGQY LFKCLINVKK EVDDALVEMH WVE Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 66%.
METT10D is a probable methyltransferase.
Specifications
| METT10D | |
| Polyclonal | |
| Unconjugated | |
| METTL16 | |
| 2610100D03Rik; 2810013M15Rik; A830095F14Rik; AI846133; methyltransferase 10 domain containing; methyltransferase 10 domain-containing protein; methyltransferase like 16; methyltransferase-like protein 16; METT10D; METTL16; N6-adenosine-methyltransferase METTL16; putative methyltransferase METT10D; RNA N6-adenosine-methyltransferase METTL16; U6 small nuclear RNA (adenine-(43)-N(6))-methyltransferase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79066 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.15 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86W50 | |
| METTL16 | |
| Recombinant protein corresponding to Human METTL16. Recombinant protein control fragment (Product #RP-91738). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction