Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ METT10D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120402
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120402 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120402 Supplier Invitrogen™ Supplier No. PA5120402
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, A-549, NIH/3T3, C6, LNCaP, Rat testis. Immunogen sequence: RIQGVPKVTY TEFCQGRTMR WALAWSFYDD VTVPSPPSKR RKLEKPRKPI TFVVLASVMK ELSLKASPLR SETAEGIVVV TTWIEKILTD LKVQHKRVPC GKEEVSLFLT AIENSWIHLR RKKRERVRQL REVPRAPEDV IQALEEKKPT PKESGNSQEL ARGPQERTPC GPALREGEAA AVEGPCPSQE SLSQEENPEP TEDERSEEKG GVEVLESCQG SSNGAQDQEA SEQFGSPVAE RGKRLPGVAG QYLFKCLINV KKEVDDALVE MHWVEGQNRD LMNQLCTYIR NQIFRLVAVN.

METT10D is a probable methyltransferase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen METT10D
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene METTL16
Gene Accession No. Q86W50, Q9CQG2
Gene Alias 2610100D03Rik; 2810013M15Rik; A830095F14Rik; AI846133; methyltransferase 10 domain containing; methyltransferase 10 domain-containing protein; methyltransferase like 16; methyltransferase-like protein 16; METT10D; METTL16; N6-adenosine-methyltransferase METTL16; putative methyltransferase METT10D; RNA N6-adenosine-methyltransferase METTL16; U6 small nuclear RNA (adenine-(43)-N(6))-methyltransferase
Gene Symbols METTL16
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360568, 67493, 79066
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.