Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
METT5D1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | METT5D1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15502320
![]() |
Novus Biologicals
NBP15502320UL |
20 μL |
Each for $206.00
|
|
|||||
NBP155023
![]() |
Novus Biologicals
NBP155023 |
100 μL |
Each for $487.50
|
|
|||||
Description
METT5D1 Polyclonal specifically detects METT5D1 in Human samples. It is validated for Western Blot.Specifications
METT5D1 | |
Polyclonal | |
Rabbit | |
Human | |
EC 2.1.1.-, FLJ33979, methyltransferase 5 domain containing 1, Methyltransferase 5 domain-containing protein 1, methyltransferase like 15, METT5D1, probable S-adenosyl-L-methionine-dependent methyltransferase METT5D1, putative S-adenosyl-L-methionine-dependent methyltransferase METT5D1 | |
METTL15 | |
IgG | |
45 kDa |
Western Blot | |
Unconjugated | |
RUO | |
A6NJ78 | |
196074 | |
Synthetic peptides corresponding to METT5D1(methyltransferase 5 domain containing 1) The peptide sequence was selected from the n terminal of METT5D1. Peptide sequence MLRYPYFCRMYKECLSCWLESGIPNLGVWPNRIHTTAEKYREYEAREQTD. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title