Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
METTL1 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299811AF594
Description
METTL1 Polyclonal specifically detects METTL1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
METTL1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
4234 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 594 | |
C12orf1, D1075-like gene product, EC 2.1.1.33, FLJ95748, methyltransferase like 1, methyltransferase-like 1, Methyltransferase-like protein 1, TRM8, tRNA (guanine-N(7)-)-methyltransferase, tRNA(m7G46)-methyltransferase, YDL201w | |
Produced in rabbits immunized with purified, recombinant METTL1 (NP_005362.3; Asp 32-Gln 265). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction