Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ METTL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580810
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human METTL1.
This gene is similar in sequence to the S. cerevisiae YDL201w gene. The gene product contains a conserved S-adenosylmethionine-binding motif and is inactivated by phosphorylation.
Specifications
METTL1 | |
Polyclonal | |
Unconjugated | |
Mettl1 | |
2810012D02Rik; C12orf1; D1075-like gene product; hypothetical protein LOC449779; methyltransferase like 1; methyltransferase-like 1; methyltransferase-like protein 1; METTL1; miRNA (guanine-N(7)-)-methyltransferase; mRNA (guanine-N(7)-)-methyltransferase; TRM8; TRMT8; tRNA (guanine(46)-N(7))-methyltransferase; tRNA (guanine-N(7)-)-methyltransferase; tRNA (guanine-N(7)-)-methyltransferase-like protein; tRNA(m7G46)-methyltransferase; Unknown (protein for MGC:134433); YDL201w; zgc:103636 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
4234 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with no preservative | |
Q9UBP6 | |
Mettl1 | |
Recombinant Human METTL1 protein (Asp32-Gln265). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction