Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
METTL10 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP321446MFV610
Description
METTL10 Polyclonal antibody specifically detects METTL10 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| METTL10 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin | |
| C10orf138, chromosome 10 open reading frame 138, EC 2.1.1.-, Em:AC068896.3, FLJ13019, methyltransferase like 10, methyltransferase-like protein 10 | |
| Produced in rabbits immunized with E. coli-derived Human METTL10 fragment | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 399818 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction