Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ METTL10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543313
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543313 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543313 Supplier Invitrogen™ Supplier No. PA543313
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SSGADGGGGA AVAARSDKGS PGEDGFVPSA LGTREHWDAV YERELQTFRE Sequence homology: Human: 100%.

Protein-lysine methyltransferase that selectively catalyzes the trimethylation of EEF1A at 'Lys-318'. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen METTL10
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene EEF1AKMT2
Gene Accession No. Q5JPI9
Gene Alias 2010208K18Rik; C10orf138; EEF1A lysine methyltransferase 2; EEF1AKMT2; Em:AC068896.3; methyltransferase like 10; methyltransferase-like protein 10; Mettl10; protein-lysine N-methyltransferase Mettl10; RGD1306300
Gene Symbols EEF1AKMT2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human LOC399818 (aa 2-51).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 399818
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.