Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTL10 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA562143
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562143 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562143 Supplier Thermo Scientific Supplier No. PA562143
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSSGADGGGG AAVAARSDKG SPGEDGFVPS ALGTREHWDA VYERELQTFR EYGDTGEIWF GEESMNRLIR WMQKHKIPL Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 26%.

METTL10 Polyclonal antibody specifically detects METTL10 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen METTL10
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EEF1AKMT2
Gene Accession No. Q5JPI9
Gene Alias 2010208K18Rik; C10orf138; EEF1A lysine methyltransferase 2; EEF1AKMT2; Em:AC068896.3; methyltransferase like 10; methyltransferase-like protein 10; Mettl10; protein-lysine N-methyltransferase Mettl10; RGD1306300
Gene Symbols EEF1AKMT2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human METTL10
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 399818
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.