Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
METTL11A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | METTL11A |
---|---|
Concentration | 0.22 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-1428
![]() ![]() |
Proteintech Group Inc
171841AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The METTL11A gene encodes an N-terminal methyltransferase for the RAN (MIM 601179) guanine nucleotide exchange factor regulator of chromosome condensation 1 (RCC1; MIM 179710). METTL11A enzyme alpha-N-methylates other protein targets such as SET (MIM 600960) and RB (MIM 180200).Specifications
METTL11A | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q5BJX0, Q8R2U4, Q9BV86 | |
28989, 362103, 66617 | |
METTL11A Fusion Protein Ag10967 | |
Primary | |
-20°C | |
NTMT1 |
0.22 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AD 003, C9orf32, methyltransferase like 11A, METTL11A, NRMT, NTM1A, NTMT1 | |
NTMT1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title