Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ METTL11B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555753
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555753 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555753 Supplier Invitrogen™ Supplier No. PA555753
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GIIILKDNVA REGCILDLSD SSVTRDMDIL RSLIRKSGLV VLGQEKQDGF PEQCIPVWMF AL Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

Alpha-N-methyltransferase that methylates the N-terminus of target proteins containing the N-terminal motif [Ala/Pro/Ser]-Pro-Lys when the initiator Met is cleaved. Specifically catalyzes monomethylation of exposed alpha-amino group of Ala or Ser residue in the [Ala/Ser]-Pro-Lys motif and Pro in the Pro-Pro-Lys motif. May activate NTMT1 by priming its substrates for trimethylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen METTL11B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Mettl11b
Gene Accession No. Q5VVY1
Gene Alias alpha N-terminal protein methyltransferase 1B; C1orf184; Gm207; HOMT1B; methyltransferase like 11B; methyltransferase-like protein 11B; METTL11B; NRMT2; NTM1B; RGD1564106; X-Pro-Lys N-terminal protein methyltransferase 1B
Gene Symbols Mettl11b
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human METTL11B. Recombinant protein control fragment (Product #RP-94569).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149281
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.