Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ METTL13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562828
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562828 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562828 Supplier Invitrogen™ Supplier No. PA562828
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LALLVVGLGG GSLPLFVHDH FPKSCIDAVE IDPSMLEVAT QWFGFSQSDR MKVHIADGLD YIASLAGGGE ARPCYDVIMF DVDSKDPTLG MSCP Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 27%.

METTL13 gene ontology annotations related to this gene include methyltransferase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen METTL13
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EEF1AKNMT
Gene Accession No. Q8N6R0
Gene Alias 5630401D24Rik; antiapoptotic protein FEAT; CGI-01; eEF1A lysine and N-terminal methyltransferase; eEF1A lysine methyltransferase; eEF1A N-terminal methyltransferase; EEF1AKNMT; eEF1A-KNMT; feat; KIAA0859; methyltransferase like 13; methyltransferase-like protein 13; METTL13; RGD1311526
Gene Symbols EEF1AKNMT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human METTL13. Recombinant protein control fragment (Product #RP-103165).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51603
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.