Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTL14 Antibody (CL4254), Novus Biologicals™
SDP

Catalog No. p-200081118 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259044 100 μL
NB403055 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP259044 Supplier Novus Biologicals Supplier No. NBP259044
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

METTL14 Monoclonal specifically detects METTL14 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen METTL14
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias EC 2.1.1, EC 2.1.1.-, KIAA1627methyltransferase-like protein 14, methyltransferase like 14
Gene Symbols METTL14
Host Species Mouse
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RSWNMDSRLQEIRERQKLRRQLLAQQLGAESADSIGAVLNSKDEQREIAETRETCRASYDTSAPNAKRKYLDEGETDEDKMEEYKDELEMQQDEE
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 57721
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.