Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ METTL14 Monoclonal Antibody (CL4254)
GREENER_CHOICE

Catalog No. PIMA524707
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524707 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524707 Supplier Invitrogen™ Supplier No. MA524707
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: RSWNMDSRLQE IRERQKLRRQ LLAQQLGAES ADSIGAVLNS KDEQREIAET RETCRASYDT SAPNAKRKYL DEGETDEDKM EEYKDELEMQ QDEE Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.

KIAA1627 is a probable methyltransferase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen METTL14
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone CL4254
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene METTL14
Gene Accession No. Q9HCE5
Gene Alias G430022H21Rik; hMETTL14; hypothetical protein zgc:77296; KIAA1627; methyltransferase like 14; methyltransferase-like protein 14; methyltransferase-like protein 14; N6-adenosine-methyltransferase subunit METTL14; Mettl14; mKIAA1627; N6-adenosine-methyltransferase non-catalytic subunit; N6-adenosine-methyltransferase subunit METTL14; RGD1304822; snoRNA MBII-84; zgc:77296
Gene Symbols METTL14
Host Species Mouse
Immunogen Recombinant protein fragment of Human METTL14 (Product #RP-96679)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57721
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.