Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ METTL14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543606
Description
Peptide sequence: LNSKDEQREI AETRETCRAS YDTSAPNAKR KYLDEGETDE DKMEEYKDEL Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
KIAA1627 is a probable methyltransferase.
Specifications
METTL14 | |
Polyclonal | |
Unconjugated | |
METTL14 | |
G430022H21Rik; hMETTL14; hypothetical protein zgc:77296; KIAA1627; methyltransferase like 14; methyltransferase-like protein 14; methyltransferase-like protein 14; N6-adenosine-methyltransferase subunit METTL14; Mettl14; mKIAA1627; N6-adenosine-methyltransferase non-catalytic subunit; N6-adenosine-methyltransferase subunit METTL14; RGD1304822; snoRNA MBII-84; zgc:77296 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
57721 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9HCE5 | |
METTL14 | |
Synthetic peptide directed towards the N-terminal of human KIAA1627 (aa 35-84). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction