Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettl14 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984118
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4118 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4118 Supplier Bioss Supplier No. BS17608R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KIAA1627 is a probable methyltransferase.

Specifications

Antigen Mettl14
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene METTL14
Gene Accession No. Q9HCE5
Gene Alias BOS_6355; G430022H21Rik; hMETTL14; hypothetical protein zgc:77296; KIAA1627; methyltransferase like 14; Methyltransferase-like protein 14; METTL14; mKIAA1627; N6-adenosine-methyltransferase non-catalytic subunit; N6-adenosine-methyltransferase subunit METTL14; RGD1304822; snoRNA MBII-84; zgc:77296
Gene Symbols METTL14
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse Mettl14.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57721
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.