Learn More
Invitrogen™ METTL17 Polyclonal Antibody

Description
METTL17 Polyclonal Antibody for Flow, IHC (P), Western Blot
Specifications
Specifications
| Antigen | METTL17 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | METTL17 |
| Gene Accession No. | Q9H7H0 |
| Gene Alias | False p73 target gene protein; methyltransferase 11 domain containing 1; methyltransferase 11 domain-containing protein 1; methyltransferase like 17; methyltransferase-like protein 17, mitochondrial; METT11D1; METTL17; Protein RSM22 homolog, mitochondrial |
| Gene Symbols | METTL17 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.