Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ METTL20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596594
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596594 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596594 Supplier Invitrogen™ Supplier No. PA596594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RYLLDNPDVV RGKSVLDLGS GCGATAIAAK MSGASRILAN DIDPIAGMAI TLNCELNRLN PFPILIQNIL NLEQDKWDLV VLGDMFYDED LADSLHQWLK KCFWTYRTRV LIGDPGRPQF SGHSIQHHLH KVVEYSLLES TRQENSGLTT STVWGFQP; Positive Samples: Mouse heart; Cellular Location: Cytoplasm, Mitochondrion matrix.

TRUSTED_SUSTAINABILITY

Specifications

Antigen METTL20
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ETFBKMT
Gene Accession No. Q6P7Q0, Q80ZM3
Gene Alias 4833442J19Rik; AI256744; C12orf72; Electron transfer flavoprotein beta subunit lysine methyltransferase; ETFB lysine methyltransferase; ETFBKMT; ETFB-KMT; methyltransferase like 20; methyltransferase-like protein 20; METTL20; Protein N-lysine methyltransferase METTL20
Gene Symbols ETFBKMT
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 105-262 of human METTL20 (NP_0011293351).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 316976, 320204
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.