Learn More
Invitrogen™ METTL21C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587871
Description
Immunogen sequence: MDVCLSSAQQ PGRRGEGLSS PGGWLEAEKK GAPQKDSTGG VLEESNKIEP SLHSLQKFVP TDYASYTQEH YRFAGKEIVI QESIESYGAV VWPGAMALCQ YLEEHAEELN FQDAKILEIG AGPGLVSIVA SILGAQVTAT DLPDVLGNLQ YNLLKNTLQC TAHLPEVKEL VWGEDLDKNF PKSAFYYDYV LASDVVYHHY FLDKLLTTMV YLSQPGTVLL WANKFRFSTD YEFLDKFKQV FDTTLLAEYP ESSVKLFKGI LKWD; Positive Samples: Mouse skeletal muscle, Mouse pancreas, Rat skeletal muscle; Cellular Location: Nucleus.
METTL21C is a protein-lysine methyltransferase.
Specifications
METTL21C | |
Polyclonal | |
Unconjugated | |
Mettl21c | |
A530098C11Rik; C13orf39; methyltransferase like 21C; methyltransferase-like protein 21C; METTL21C; Protein-lysine methyltransferase METTL21C; RGD1561425 | |
Rabbit | |
Affinity chromatography | |
RUO | |
301378, 433294 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BLU2 | |
Mettl21c | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-264 of human METTL21C (NP_0010109771). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.