Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ METTL22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567334
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567334 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567334 Supplier Invitrogen™ Supplier No. PA567334
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NACTAILSVEK RLNFTLRHLD VTCEAYDHFR SCLHALEQLT DGKLRFVVEP VEASFPQLLV YERLQQLELW KIIAEP Highest antigen sequence identity to the following orthologs - mouse 82%, rat 82%.

Protein N-lysine methyltransferase. In vitro methylates KIN.
TRUSTED_SUSTAINABILITY

Specifications

Antigen METTL22
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene METTL22
Gene Accession No. Q9BUU2
Gene Alias C16orf68; C25H16orf68; LP8272; methyltransferase like 22; methyltransferase-like protein 22; Mettl22; RGD1311433
Gene Symbols METTL22
Host Species Rabbit
Immunogen Recombinant Human METTL22. Recombinant protein control fragment (Product #RP-107982).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79091
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.