Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ METTL22 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569561
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 79%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 93%.
Protein N-lysine methyltransferase. In vitro methylates KIN.
Specifications
METTL22 | |
Polyclonal | |
Unconjugated | |
METTL22 | |
C16orf68; C25H16orf68; LP8272; methyltransferase like 22; methyltransferase-like protein 22; Mettl22; RGD1311433 | |
Rabbit | |
Affinity chromatography | |
RUO | |
79091 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BUU2 | |
METTL22 | |
synthetic peptide directed towards the C-terminal region of human METTL22. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction