Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
METTL25 Antibody (OTI2B6), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272656IR
Description
METTL25 Monoclonal specifically detects METTL25 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| METTL25 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| METTL25 | |
| Full length recombinant protein of human METTL25 (NP_115606) produced in HEK293T cells. | |
| 0.1 mL | |
| 84190.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI2B6 | |
| Western Blot, Immunohistochemistry | |
| C12orf26, chromosome 12 open reading frame 26, FLJ22789, hypothetical protein LOC84190 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction