Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ METTL2B Monoclonal Antibody (2A9)
GREENER_CHOICE

Catalog No. PIMA522201
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522201 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522201 Supplier Invitrogen™ Supplier No. MA522201
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: SQNQNHLKDW FLENKSEVCE CRNNEDGPGL IMEEQHKCSS KSLEHKTQTP PVEENVTQKI SDLEICAD.

This gene is a member of a family of methyltransferases that share homology with, but are distinct from, the UbiE family of methyltransferases. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen METTL2B
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2A9
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene METTL2B
Gene Accession No. Q6P1Q9
Gene Alias methyltransferase like 2B; methyltransferase-like protein 2B; METL; METTL2; METTL2A; METTL2B; PSENIP1; tRNA N(3)-methylcytidine methyltransferase METTL2B
Gene Symbols METTL2B
Host Species Mouse
Immunogen METTL2B (NP_060866.1, 41 a.a. ~ 108 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55798
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.