Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ METTL2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531303
Description
Recommended positive controls: U87-MG, IMR32. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene is a member of a family of methyltransferases that share homology with, but are distinct from, the UbiE family of methyltransferases. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
Specifications
| METTL2B | |
| Polyclonal | |
| Unconjugated | |
| METTL2B | |
| methyltransferase like 2B; methyltransferase-like protein 2B; METL; METTL2; METTL2A; METTL2B; PSENIP1; tRNA N(3)-methylcytidine methyltransferase METTL2B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55798 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q6P1Q9 | |
| METTL2B | |
| Recombinant protein encompassing a sequence within the center region of human METTL2B. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction