Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTL3 Monoclonal Antibody (29C8), Active Motif™
SDP

Catalog No. 502277272
Encompass
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7272 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7272 Supplier Active Motif Supplier No. 65688
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

METTL3 Monoclonal antibody specifically detects METTL3 in Human samples. It is validated for Immunoprecipitation, Western Blot

Specifications

Antigen METTL3
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 29C8
Conjugate Unconjugated
Formulation tissue culture supernatant with 30% glycerol and 0.035% sodium azide
Gene METTL3
Gene Accession No. Q86U44
Gene Alias 2310024F18Rik; adoMet-binding subunit of the human mRNA (N6-adenosine)-methyltransferase; fj82g07; hMETTL3; IME4; M6A; m6a methyltransferase; methyltransferase like 3; methyltransferase-like 3; methyltransferase-like protein 3; METTL3; mRNA (2'-O-methyladenosine-N(6)-)-methyltransferase; mRNA m(6)A methyltransferase; MTA70; MT-A70; N6-adenosine-methyltransferase 70 kDa subunit; N6-adenosine-methyltransferase catalytic subunit; N6-adenosine-methyltransferase subunit METTL3; Spo8; Unknown (protein for MGC:77093); wu:fj82g07; zgc:77093
Host Species Mouse
Immunogen This antibody was raised against a peptide within the N-terminal region of human METTL3 (N6-adenosine-methyltransferase).
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56339
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.