Learn More
METTL3/MT-A70 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | METTL3/MT-A70 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1930
![]() |
Bethyl Laboratories, Inc
IHC00342T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1929
![]() |
Bethyl Laboratories, Inc
IHC00342 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan, Monkey, Gorilla,
METTL3 is the 70 kDa subunit of MT-A which is part of N6-adenosine-methyltransferase. This enzyme is involved in the posttranscriptional methylation of internal adenosine residues in eukaryotic mRNAs, forming N6-methyladenosine.Specifications
METTL3/MT-A70 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
IME4, M6A, MT-A70, Spo8, N6-adenosine-methyltransferase 70 kDa subunit, adoMet-binding subunit of the human mRNA (N6-adenosine)-methyltransferase, mRNA m(6)A methyltransferase, methyltransferase-like protein 3 | |
Between 1 and 50 | |
Primary | |
4° C | |
METTL3 |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q86U44, Q8C3P7 | |
56335, 56339 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.