Learn More
METTL3/MT-A70 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301567A
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
METTL3 is the 70 kDa subunit of MT-A which is part of N6-adenosine-methyltransferase. This enzyme is involved in the posttranscriptional methylation of internal adenosine residues in eukaryotic mRNAs, forming N6-methyladenosine.Specifications
| METTL3/MT-A70 | |
| Polyclonal | |
| Unconjugated | |
| METTL3 | |
| IME4, M6A, MT-A70, Spo8, N6-adenosine-methyltransferase 70 kDa subunit, adoMet-binding subunit of the human mRNA (N6-adenosine)-methyltransferase, mRNA m(6)A methyltransferase, methyltransferase-like protein 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56335, 56339 | |
| 4° C | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q86U44, Q8C3P7 | |
| METTL3 | |
| Between 1 and 50 | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.