Learn More
Invitrogen™ METTL3 Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (80%), Xenopus laevis (81%), Chimpanzee (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | METTL3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.54 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | METTL3 |
| Gene Accession No. | Q86U44 |
| Gene Alias | 2310024F18Rik; adoMet-binding subunit of the human mRNA (N6-adenosine)-methyltransferase; fj82g07; hMETTL3; IME4; M6A; m6a methyltransferase; methyltransferase like 3; methyltransferase-like 3; methyltransferase-like protein 3; METTL3; mRNA (2'-O-methyladenosine-N(6)-)-methyltransferase; mRNA m(6)A methyltransferase; MTA70; MT-A70; N6-adenosine-methyltransferase 70 kDa subunit; N6-adenosine-methyltransferase catalytic subunit; N6-adenosine-methyltransferase subunit METTL3; Spo8; Unknown (protein for MGC:77093); wu:fj82g07; zgc:77093 |
| Gene Symbols | METTL3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.