Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ METTL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121190
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121190 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121190 Supplier Invitrogen™ Supplier No. PA5121190
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T, HT-1080, NIH/3T3, Mouse testis, Mouse brain, Rat spleen. The target is usually found in the following locations: Nucleus speckle. Immunogen sequence: MSDTWSSIQA HKKQLDSLRE RLQRRRKQDS GHLDLRNPEA ALSPTFRSDS PVPTAPTSGG PKPSTASAVP ELATDPELEK KLLHHLSDLA LTLPTDAVSI CLAISTPDAP ATQDGVESLL QKFAAQELIE VKRGLLQDDA HPTLVTYADH SKLSAM.

METTL3 is the 70 kDa subunit of MT-A which is part of N6-adenosine-methyltransferase. This enzyme is involved in the posttranscriptional methylation of internal adenosine residues in eukaryotic mRNAs, forming N6-methyladenosine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen METTL3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene METTL3
Gene Accession No. Q86U44, Q8C3P7
Gene Alias 2310024F18Rik; adoMet-binding subunit of the human mRNA (N6-adenosine)-methyltransferase; fj82g07; hMETTL3; IME4; M6A; m6a methyltransferase; methyltransferase like 3; methyltransferase-like 3; methyltransferase-like protein 3; METTL3; mRNA (2'-O-methyladenosine-N(6)-)-methyltransferase; mRNA m(6)A methyltransferase; MTA70; MT-A70; N6-adenosine-methyltransferase 70 kDa subunit; N6-adenosine-methyltransferase catalytic subunit; N6-adenosine-methyltransferase subunit METTL3; Spo8; Unknown (protein for MGC:77093); wu:fj82g07; zgc:77093
Gene Symbols METTL3
Host Species Rabbit
Immunogen Recombinant Protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361035, 56335, 56339
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.