Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ METTL4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA523881
Description
METTL4 Polyclonal Antibody for Western Blot, IHC (P), Flow
METTL4 gene ontology annotations related to this gene include methyltransferase activity; nucleic acid binding.
Specifications
METTL4 | |
Polyclonal | |
Unconjugated | |
METTL4 | |
2410198H06Rik; A730091E08Rik; AV296509; HsT661; methyltransferase like 4; methyltransferase-like protein 4; METTL4; N(6)-adenine-specific DNA methyltransferase METTL4; N(6)-adenine-specific methyltransferase METTL4; N(6)-adenine-specific methyltransferase METTL4; methyltransferase-like protein 4; RGD1306451 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
316731, 64863 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.48 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8N3J2 | |
METTL4 | |
KLH conjugated synthetic peptide between 315-344 amino acids from the C-terminal region of human METTL4. | |
200 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction