Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ METTL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111243
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111243 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111243 Supplier Invitrogen™ Supplier No. PA5111243
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IIGKKRKRCV VFNQGELDAM EYHTKIRELI LDGSLQLIQE GLKSGFLYPL FEKQDKGSKP ITLPLDACSL SELCEMAKHL PSLNEMEHQT LQLVEEDTSV TEQDLFLRVV ENNSSFTK Highest antigen sequence indentity to the following orthologs: Rat - 77%, Mouse - 78%.

METTL4 gene ontology annotations related to this gene include methyltransferase activity; nucleic acid binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen METTL4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene METTL4
Gene Accession No. Q8N3J2
Gene Alias 2410198H06Rik; A730091E08Rik; AV296509; HsT661; methyltransferase like 4; methyltransferase-like protein 4; METTL4; N(6)-adenine-specific DNA methyltransferase METTL4; N(6)-adenine-specific methyltransferase METTL4; N(6)-adenine-specific methyltransferase METTL4; methyltransferase-like protein 4; RGD1306451
Gene Symbols METTL4
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human METTL4. Recombinant protein control fragment (Product #RP-108376).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64863
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.