Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ METTL6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557185
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557185 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557185 Supplier Invitrogen™ Supplier No. PA557185
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MASLQRKGLQ ARILTSEEEE KLKRDQTLVS DFKQQKLEQE AQKNWDLFYK RNSTNFFKDR HWTTREFEEL RSCREFEDQ Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 86%.

S-adenosyl-L-methionine-dependent methyltransferase that mediates N(3)-methylcytidine modification of residue 32 of the tRNA anticodon loop of tRNA(Ser). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen METTL6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene METTL6
Gene Accession No. Q8TCB7
Gene Alias 1600013P15Rik; AI467521; AU020711; methyltransferase like 6; methyltransferase-like protein 6; Mettl6; tRNA N(3)-methylcytidine methyltransferase METTL6
Gene Symbols METTL6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human METTL6. Recombinant protein control fragment (Product #RP-96490).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131965
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.