Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTL7A Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP177336

Catalog No. NBP177336


Only null left
Add to Cart

Description

Description

METTL7A Polyclonal specifically detects METTL7A in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

METTL7A
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
AAM-B, DKFZP586A0522, EC 2.1.1, EC 2.1.1.-, methyltransferase like 7A, methyltransferase-like protein 7A, Protein AAM-B
Rabbit
Affinity Purified
RUO
25840
Human, Mouse
IgG
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
1 mg/mL
Western Blot 2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2 μg/mL
NP_054752
METTL7A
Antibody was raised against a 13 amino acid synthetic peptide near the center of human MettL7A. The immunogen is located within amino acids 130 - 180 of MettL7A. Amino Acid Squence: SVKNQERILREV
0.1 mg
Primary
At least two isoforms of MettL7A are known to exist. This antibody is predicted to not cross-react with MettL7B.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only